Zip bags, double pocket without writing area, Corning® Gosselin™
Toimittaja: Corning
SP2-18 SP8-01 SP2-01
GOSSSP2-18EA 0 EUR
GOSSSP2-18 GOSSSP8-01 GOSSSP2-01
Zip bags, double pocket without writing area, Corning® Gosselin™
Pussit ja sulkijat Näytepussit
Double pocket ZIP polyethylene bags.
  • Non sterile
  • Quick closure system for efficient transport of samples