Bottles, narrow neck, square, HDPE, with screw caps, Corning® Gosselin™
Toimittaja: Corning
FC1000-52 FC1000-51 FC1000-50
GOSSFC1000-52EA 242 EUR
GOSSFC1000-52 GOSSFC1000-51 GOSSFC1000-50
Bottles, narrow neck, square, HDPE, with screw caps, Corning® Gosselin™
Pullot Muovipullot
Made of HDPE, translucent, with PP screw cap.
  • Graduated
  • Good chemical and temperature resistance
  • Tamper evident screw cap with shaped seal
  • Screw cap assembled