Racks for upright freezers, slide boxes
Toimittaja: GLW
K303 K304 K305 K306 K307 K308 K309
GLWAK303EA 126 EUR
GLWAK303 GLWAK304 GLWAK305 GLWAK306 GLWAK307 GLWAK308 GLWAK309
Racks for upright freezers, slide boxes
Koeputkitelineet Telineet kryolaatikoille
These freezer racks are made of stainless steel that are used for all common upright freezers available in different heights.
  • Drawers can hold upto 3 to 6 slide boxes
  • Each drawer is easily removable and has an ergonomic handle