Cryoboxes B10/B12/B20
Toimittaja: GLW
B20 B20B B20G B20R B20Y B10 B10B B10G B10R B10Y B10S BS10 B12 B12B B12G B12R B12Y B12S BS12 B12BS B12GS B12RS B12YS B20BS B20GS B20RS B20S B20YS BS20
GLWAB20EA 8.6 EUR
GLWAB20 GLWAB20B GLWAB20G GLWAB20R GLWAB20Y GLWAB10 GLWAB10B GLWAB10G GLWAB10R GLWAB10Y GLWAB10S GLWABS10 GLWAB12 GLWAB12B GLWAB12G GLWAB12R GLWAB12Y GLWAB12S GLWABS12 GLWAB12BS GLWAB12GS GLWAB12RS GLWAB12YS GLWAB20BS GLWAB20GS GLWAB20RS GLWAB20S GLWAB20YS GLWABS20
Cryoboxes B10/B12/B20
Laatikot Kryogeenipullolaatikot
Cryoboxes, made of PP, natural or coloured.
  • Autoclavable up to +121 °C and use in freezer down to -90 °C
  • Included clear lid offers a view of the contents without opening the cryobox
  • Drainage holes in base for easy opening the box at low temperatures
  • For light sensitive samples please choose the color BLACK/black
Suitable for 10 tubes up to 30 mm diameter and 2 tubes up to 17 mm diameter.
B10-Series:
Designed for 20 ml up to 30 ml scintillation vials and culture counter tubes. Generic tube size up to 80 mm height and 30 mm diameter.
B20-Series:
More Product Information