Serological pipettes
Toimittaja: GILSON
F110121 F110123 F110125 F110127 F110129 F110122 F110124 F110126 F110128 F110130 F110131
GILSF110121EA 136.4 EUR
GILSF110121 GILSF110123 GILSF110125 GILSF110127 GILSF110129 GILSF110122 GILSF110124 GILSF110126 GILSF110128 GILSF110130 GILSF110131
Serological pipettes
Pipetit ja pipetintäyttäjät Serologiset pipetit
The Gilson disposable plastic serological pipettes are perfect for use with MACROMAN® to help you in your daily cell-culture, molecular biology and microbiology applications. Ideal for scientists who need to easily and quickly transfer liquid aliquots from one container to another with precision.
  • USP Class VI polystyrene and are free of contaminants that can alter your experiment
  • Gamma-sterilised, non-pyrogenic, and certified free of detectable RNase/DNase
  • Polyester fiber plug, offering a high level of protection against over-pipetting and contamination by