Fraction collectors
Toimittaja: GILSON LCMS
171011 171011DV 171041
GILI171011EA 3910 EUR
GILI171011 GILI171011DV GILI171041
Fraction collectors
Fraktionkerääjät
Ideal solutions for small to large capacity collection. Gilson offers advanced solutions for fraction collection with a complete line of stand‑alone and software-controlled fraction collectors ranging from small footprint to large vessel capacity models. The built-in, easy-to-use keypad makes setup and operation simple and capable of interfacing with any HPLC system or control the system through your PC with TRILUTION® LC software.
  • Collect fractions by time, drop, or peak to suit all your LC applications
  • Choose the compact FC 203B for small-capacity