Ergonomic computer mouse, GCQ® EasyMouse
Toimittaja: GETT GERATETECHNIK
KH29210 KH29211
GETTKH29210EA 48.7 EUR
GETTKH29210 GETTKH29211
Ergonomic computer mouse, GCQ® EasyMouse
Tietokoneen hiiret ja hiirimatot
The EasyMouse is an ergonomic and reliable plastic mouse. It is equipped with two mouse buttons and a scroll wheel.
  • Medical grade IP 68 mouse
  • Completely waterproof and dustproof
  • Easy to clean and disinfect
  • Silicone cap protects USB connector from splashing water during cleaning
  • Ergonomic functionality with two mouse buttons and a scroll wheel
  • Symmetrical housing shape is suitable for right- and left-handed users
  • Optical sensor
  • Resolution: 1000 dpi
  • Button life >=2 million actuations
  • Engineered in Germany
The EasyMouse is an ergonomic and reliableMore Product Information