Silicone encased keyboard for hygienic environments, InduProof® Smart Touch
Toimittaja: GETT GERATETECHNIK
KG23221 KG24209 KG25200 KG25208
GETTKG23221EA 291 EUR
GETTKG23221 GETTKG24209 GETTKG25200 GETTKG25208
Silicone encased keyboard for hygienic environments, InduProof® Smart Touch
Näppäimistöt
Fully waterproof and dustproof medical keyboard with number block and touchpad.
  • Dustproof and waterproof keyboard with integrated capacitive touchpad
  • Can be completely disinfected
  • Soft typing through silicone switching elements
  • Flat key caps for thorough disinfection
  • Pleasant tactile feeling even with protective gloves
  • Three year warranty on technical function
  • Made in Germany
This InduProof® keyboard has a complete set of keys with numeric keypad, an integrated touchpad, and provides a pleasant tactile feel and the highest degree of protection. ThisMore Product Information