Medical keyboards, Induproof® smart compact
Toimittaja: GETT GERATETECHNIK
KG20230 KG20231 KG21247 KG23252 KG21204 KG23253 KG21226 KG21223 KG22304 KG24247
GETTKG20230EA 180 EUR
GETTKG20230 GETTKG20231 GETTKG21247 GETTKG23252 GETTKG21204 GETTKG23253 GETTKG21226 GETTKG21223 GETTKG22304 GETTKG24247
Medical keyboards, Induproof® smart compact
Näppäimistöt
Compact silicone keyboard with 86 keys including num pad for use in cramped working environments.
  • Ergonomic design
  • White housing colour
  • Complies with USB 2.0
  • Flat key profile for easy cleaning
  • Comfortable typing
  • IP68 protection, can be submerged into liquids
Keylock function to disable the keys while cleaning the keyboard.
Can be cleaned with soapy water, disinfectants or other cleaning agents.
Sertifikaatit: CE, FCC certified.