Silicone keyboards with integrated mouse button, InduProof® Advanced MB
Toimittaja: GETT GERATETECHNIK
KG19259 KG21212
GETTKG19259EA 293 EUR
GETTKG19259 GETTKG21212
Silicone keyboards with integrated mouse button, InduProof® Advanced MB
Näppäimistöt
Rugged, waterproof and dustproof silicone keyboard with integrated mouse button.
  • Rugged, durable keyboard with 104 keys and integrated mouse button
  • Comfortable and ergonomic operation, even with protective gloves (silicone or latex)
  • IP 68 protection; device can be submerged into liquids
  • Easy cleaning and disinfection, even under tap water or by immersion
  • Increased chemical and mechanical resistance due to special surface treatment
  • VESA-M5-boreholes (distance 75×75mm)
  • 4-year warranty on technical function
  • Made in Germany
The InduProof® Advanced has aMore Product Information