Precision hotplates with separate controller
Toimittaja: GESTIGKEIT HARRY
PZ35EB
GESTPZ35EBEA 2250 EUR
GESTPZ35EB
Precision hotplates with separate controller
Lämmittimet Lämpölevyt
This precision hotplate is equipped with adjustable temperature sensors (100 to 350 °C). These sensors function as an emergency control which protects the appliance and the substance being heated from dangerous overheating.