Aluminium foils, round
Toimittaja: GERBER FUNKE
5712
GERB5712EA 131 EUR
GERB5712
Aluminium foils, round
Alumiinifoliot
These aluminium foils are used with reference drier RD-8 for determining the moisture content of powdered milk.