Affinity chromatography columns, HiTrap™ IXSelect
Toimittaja: Cytiva
17-3714-11 17-3714-12
GEHE17-3714-11EA 1052 EUR
GEHE17-3714-11 GEHE17-3714-12
Affinity chromatography columns, HiTrap™ IXSelect
Kromatografiakolonnit
HiTrap™ IXSelect columns are prepacked with IXSelect resin for screening and small-scale protein purifications using affinity chromatography.
  • IXSelect is a high-throughput resin specifically designed for the purification of factor IX targets from recombinant feed streams
Columns for screening of selectivity, binding and elution conditions, as well as small scale purifications.