Cryogenic Colour Dots Writable Labels
Toimittaja: GA
LT-11A LT-11BE LT-11BL LT-11BR LT-11GA LT-11GS LT-11LA LT-11LE LT-11MT LT-11OR LT-11PI LT-11RE LT-11SI LT-11VI LT-11WH LT-11X7A LT-11YE LT-13A LT-13BE LT-13BL LT-13BR LT-13GA LT-13GS LT-13LA LT-13LE LT-13MT LT-13OR LT-13PI LT-13RE LT-13SI LT-13VI LT-13WH LT-13YE LT-19BL LT-19GS LT-19PI LT-19RE LT-19WH LT-19YE LT-9A LT-9BE LT-9BL LT-9BR LT-9GA LT-9GS LT-9LA LT-9LE LT-9OR LT-9PI LT-9RE LT-9SI LT-9VI LT-9WH LT-9X8A LT-9YE
GAINLT-11AEA 34.7 EUR
GAINLT-11A GAINLT-11BE GAINLT-11BL GAINLT-11BR GAINLT-11GA GAINLT-11GS GAINLT-11LA GAINLT-11LE GAINLT-11MT GAINLT-11OR GAINLT-11PI GAINLT-11RE GAINLT-11SI GAINLT-11VI GAINLT-11WH GAINLT-11X7A GAINLT-11YE GAINLT-13A GAINLT-13BE GAINLT-13BL GAINLT-13BR GAINLT-13GA GAINLT-13GS GAINLT-13LA GAINLT-13LE GAINLT-13MT GAINLT-13OR GAINLT-13PI GAINLT-13RE GAINLT-13SI GAINLT-13VI GAINLT-13WH GAINLT-13YE GAINLT-19BL GAINLT-19GS GAINLT-19PI GAINLT-19RE GAINLT-19WH GAINLT-19YE GAINLT-9A GAINLT-9BE GAINLT-9BL GAINLT-9BR GAINLT-9GA GAINLT-9GS GAINLT-9LA GAINLT-9LE GAINLT-9OR GAINLT-9PI GAINLT-9RE GAINLT-9SI GAINLT-9VI GAINLT-9WH GAINLT-9X8A GAINLT-9YE
Cryogenic Colour Dots Writable Labels
Etiketit, tarrat
These small diameter, round cryo labels for microtube tops are made with a strong adhesive. Ideal for −80 °C freezers, dry ice, and liquid nitrogen storage.
  • Temperature range: −196 to 110 °C (−321 to 230 °F)
  • Available in a wide range of colours, for colour-coding and easy identification
  • Easy to write on with permanent markers
  • Small round labels are ideal for 0,5 to 1,5 ml Eppendorf tube caps and vials
  • Strong adhesive ensures labels won't wrinkle or peel during long-term storage
  • Resistant to gamma irradiation (tested to up to 50 kGy)
Specially designedMore Product Information