Rectangle and Circle Cryogenic Labels for Thermal Transfer Barcode Printers
Toimittaja: GA
JTTA-103C1-2WH JTTA-158C1-2WH JTTA-161C1-2WH JTTA-172C1-2WH JTTA-172C3-4WH JTTA-183C1-2WH JTTA-228C1-2WH JTTA-274C1-2WH JTTA52NOTC12WH JTTA52NOTC34WH JTTA-70C1-2WH JTTA-70C3-4WH
GAINJTTA-103C1-2WHEA 105 EUR
GAINJTTA-103C1-2WH GAINJTTA-158C1-2WH GAINJTTA-161C1-2WH GAINJTTA-172C1-2WH GAINJTTA-172C3-4WH GAINJTTA-183C1-2WH GAINJTTA-228C1-2WH GAINJTTA-274C1-2WH GAINJTTA52NOTC12WH GAINJTTA52NOTC34WH GAINJTTA-70C1-2WH GAINJTTA-70C3-4WH
Rectangle and Circle Cryogenic Labels for Thermal Transfer Barcode Printers
Etiketit, tarrat
These labels are available in a range of sizes and configurations, perfect for identifying cryo vials, microtubes, cardboard or polypropylene cryo/freezer boxes, cell culture plates, and plastic bags.
  • Temperature range: −196 to +110 °C
  • Resistant to long-term cryogenic storage in freezers and liquid nitrogen (−196 °C)
  • Resistant to spraying and swabbing with alcohol and surface sanitising wipes
  • Requires no cure time before liquid nitrogen immersion; will also adhere to wet surfaces
  • High-quality thermal-transfer printing, ideal for serialised data and