Wrap-Around Cryogenic Thermal Transfer Labels
Toimittaja: GA
HBTT303NOT11BL HBTT303NOT11GA HBTT303NOT11LA HBTT303NOT11OR HBTT303NOT11PI HBTT303NOT11RE HBTT303NOT11SI HBTT303NOT11WH HBTT303NOT11YE HBTT303NOT31BL HBTT303NOT31GA HBTT303NOT31LA HBTT303NOT31OR HBTT303NOT31PI HBTT303NOT31RE HBTT303NOT31SI HBTT303NOT31WH HBTT303NOT31YE HBTT308NOT11WH HBTT308NOT31WH HBTT309NOT11GA HBTT309NOT11PI HBTT309NOT11WH HBTT309NOT11YE HBTT309NOT31GA HBTT309NOT31PI HBTT309NOT31WH HBTT309NOT31YE HBTT311NOT12WH HBTT311NOT34WH HBTT312NOT11WH HBTT312NOT33WH HBTT312NPC11WH HBTT312NPC33WH HBTT313C1-2WH HBTT313C3-2WH HBTT313SBC12WH HBTT313SBC32WH HBTT314C1-2WH HBTT314C3-2WH HBTT317NOT11WH HBTT317NOT32WH HBTT322NOT12WH HBTT322NOT34WH HBTT324C10.5WH HBTT324NOT31WH HBTT337NPC11WH HBTT337NPC31WH HBTT338NPC11WH HBTT338NPC32WH HBTT339NPC11BL HBTT339NPC11GA HBTT339NPC11LA HBTT339NPC11OR HBTT339NPC11SI HBTT339NPC11WH HBTT339NPC11YE HBTT339NPC32BL HBTT339NPC32GA HBTT339NPC32LA HBTT339NPC32OR HBTT339NPC32SI HBTT339NPC32WH HBTT339NPC32YE HBTT340C12.5GA HBTT340C125LBL HBTT340C12.5RE HBTT340C12.5WH HBTT340NOT35GA HBTT340NO35LBL HBTT340NOT35RE HBTT340NOT35WH HBTT341NOT11WH HBTT341NOT31WH HBTT342NPC11WH HBTT342NPC31WH HBTT343NOT11WH HBTT343NOT31WH HBTT344NOT11WH HBTT344NOT32WH HBTT346NPC11WH HBTT346NPC32WH
GAINHBTT303NOT11BLEA 89.1 EUR
GAINHBTT303NOT11BL GAINHBTT303NOT11GA GAINHBTT303NOT11LA GAINHBTT303NOT11OR GAINHBTT303NOT11PI GAINHBTT303NOT11RE GAINHBTT303NOT11SI GAINHBTT303NOT11WH GAINHBTT303NOT11YE GAINHBTT303NOT31BL GAINHBTT303NOT31GA GAINHBTT303NOT31LA GAINHBTT303NOT31OR GAINHBTT303NOT31PI GAINHBTT303NOT31RE GAINHBTT303NOT31SI GAINHBTT303NOT31WH GAINHBTT303NOT31YE GAINHBTT308NOT11WH GAINHBTT308NOT31WH GAINHBTT309NOT11GA GAINHBTT309NOT11PI GAINHBTT309NOT11WH GAINHBTT309NOT11YE GAINHBTT309NOT31GA GAINHBTT309NOT31PI GAINHBTT309NOT31WH GAINHBTT309NOT31YE GAINHBTT311NOT12WH GAINHBTT311NOT34WH GAINHBTT312NOT11WH GAINHBTT312NOT33WH GAINHBTT312NPC11WH GAINHBTT312NPC33WH GAINHBTT313C1-2WH GAINHBTT313C3-2WH GAINHBTT313SBC12WH GAINHBTT313SBC32WH GAINHBTT314C1-2WH GAINHBTT314C3-2WH GAINHBTT317NOT11WH GAINHBTT317NOT32WH GAINHBTT322NOT12WH GAINHBTT322NOT34WH GAINHBTT324C10.5WH GAINHBTT324NOT31WH GAINHBTT337NPC11WH GAINHBTT337NPC31WH GAINHBTT338NPC11WH GAINHBTT338NPC32WH GAINHBTT339NPC11BL GAINHBTT339NPC11GA GAINHBTT339NPC11LA GAINHBTT339NPC11OR GAINHBTT339NPC11SI GAINHBTT339NPC11WH GAINHBTT339NPC11YE GAINHBTT339NPC32BL GAINHBTT339NPC32GA GAINHBTT339NPC32LA GAINHBTT339NPC32OR GAINHBTT339NPC32SI GAINHBTT339NPC32WH GAINHBTT339NPC32YE GAINHBTT340C12.5GA GAINHBTT340C125LBL GAINHBTT340C12.5RE GAINHBTT340C12.5WH GAINHBTT340NOT35GA GAINHBTT340NO35LBL GAINHBTT340NOT35RE GAINHBTT340NOT35WH GAINHBTT341NOT11WH GAINHBTT341NOT31WH GAINHBTT342NPC11WH GAINHBTT342NPC31WH GAINHBTT343NOT11WH GAINHBTT343NOT31WH GAINHBTT344NOT11WH GAINHBTT344NOT32WH GAINHBTT346NPC11WH GAINHBTT346NPC32WH
Wrap-Around Cryogenic Thermal Transfer Labels
Etiketit, tarrat
Transparent cryogenic self-laminating labels for thermal transfer printers. Their clear laminate protects the printed information from alcohols and detergents to ensure the secure identification of tubes and cryo vials. These labels will also withstand prolonged storage under cryogenic conditions, including in liquid phase and vapor phase liquid nitrogen.
  • Temperature range: –196 to +100 °C (–321 to +212 °F)
  • Resist cryogenic conditions storage such as vapor phase and liquid phase liquid nitrogen
  • Labels are self-laminating, protecting the information from