Thermal Transfer Cryogenic Cover-Up Labels for Frozen Surfaces
Toimittaja: GA
BUC-29SBC1-1SW BUC-29SBC3-3SW BUC-7C1-1SW BUC-7C3-5SW BUC-10C1-1SW BUC-10C3-2SW BUC-127C1-1SW BUC-127C3-2SW BUC-192C1-0.25 BUC-192C3-0.5S BUC-28C1-1SW BUC-28C3-2SW BUC-29C1-1SW BUC-29C3-3SW
GAINBUC-29SBC1-1SWEA 61.3 EUR
GAINBUC-29SBC1-1SW GAINBUC-29SBC3-3SW GAINBUC-7C1-1SW GAINBUC-7C3-5SW GAINBUC-10C1-1SW GAINBUC-10C3-2SW GAINBUC-127C1-1SW GAINBUC-127C3-2SW GAINBUC-192C1-0.25 GAINBUC-192C3-0.5S GAINBUC-28C1-1SW GAINBUC-28C3-2SW GAINBUC-29C1-1SW GAINBUC-29C3-3SW
Thermal Transfer Cryogenic Cover-Up Labels for Frozen Surfaces
Etiketit, tarrat
Blackout labels can be affixed to both frozen and ambient temperature containers in order to conceal their existing information. These thermal transfer cryo blackout labels allow cryo vials and tubes to be re-labeled without the need for thawing, cure time, or any other special treatment.
  • Temperature range: –196 to 121 °C
  • Ideal for permanently obscuring all pre-existing information
  • Permanent adhesive ensures no flagging or detachment when stored in liquid nitrogen
  • Ideal for labeling already frozen surfaces (as low as –80 °C), without thawing first
  • No