Fisher Container™ Precision ZipClean Pouches™
Toimittaja: FISHER CONTAINER
91089P 91086P 91085P 91090P 91087P 91088P
FISH91089PEA 1960 EUR
FISH91089P FISH91086P FISH91085P FISH91090P FISH91087P FISH91088P
Fisher Container™ Precision ZipClean Pouches™
Pussit ja sulkijat Säilytyspussit
Plastic bags. Tough, ultraclean, high clarity film that is ideal for all multi-use, reclosable cleanroom and laboratory applications.
  • Easy open and easy zip closure
  • Cleanroom certified
  • Sealed in a double-lined carton
  • Barrier properties
  • Suitable for sterilization by gamma irradiation
Pakkaus: All Precision ZipClean Pouches® are manufactured in strict cleanroom conditions. Double bagged and sealed in a double lined carton.