Kaliumjodidi 200 g/l in aqueous solution

Toimittaja: FF CHEMICALS

Synonyms: Dipotassium diiodide, Kali iodide

FF7091000
FFCHFF7091000EA 0 EUR
FFCHFF7091000
Kaliumjodidi 200 g/l in aqueous solution
Kaliumjodidi
Kaava: Kl
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011405
CAS-numero: 7681-11-0
EINECS: 231-659-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR