Formaldehydi 20% in aqueous solution

Toimittaja: FF CHEMICALS
Vaara

Synonyms: Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde

FF6635000
FFCHFF6635000EA 0 EUR
FFCHFF6635000
Formaldehydi 20% in aqueous solution
Formaldehydi
Kaava: CH₂O
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00003274
CAS-numero: 50-00-0
EINECS: 200-001-8
UN: 3334
ADR: 9,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR