Kaliumjodidi 3 mol/l (3 N) in aqueous solution

Toimittaja: FF CHEMICALS

Synonyms: Dipotassium diiodide, Kali iodide

FF60509010
FFCHFF60509010EA 0 EUR
FFCHFF60509010
Kaliumjodidi 3 mol/l (3 N) in aqueous solution
Kaliumjodidi
Kaava: Kl
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011405
CAS-numero: 7681-11-0
EINECS: 231-659-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR