Formalin 10% in aqueous phosphate buffer (pH 7.0 ± 0.2)

Toimittaja: FF CHEMICALS
Varoitus

FF1150035 FF1195000 FF1209010 FF22023000 FF22589200 FF8370150 FF8860080
FFCHFF1150035EA 0 EUR
FFCHFF1150035 FFCHFF1195000 FFCHFF1209010 FFCHFF22023000 FFCHFF22589200 FFCHFF8370150 FFCHFF8860080
Formalin 10% in aqueous phosphate buffer (pH 7.0 ± 0.2)
Formalin

(= 4% Formaldehyde solution)

Kaava: CH₂O
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00003274
CAS-numero: 50-00-0
EINECS: 200-001-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR