FARRAR® ULC Ultra-Low Chambers
Toimittaja: TRANE TECHNOLOGIES
ULC259-BWC50HZ ULC328-SAC50HZ ULC190-SAC50HZ ULC190-SWC50HZ ULC259-DAC50HZ ULC259-DWC50HZ ULC259-SWC50HZ ULC259-BAC50HZ ULC259-SAC50HZ ULC328-SWC50HZ
FARRULC259-BWC50HZEA 0 EUR
FARRULC259-BWC50HZ FARRULC328-SAC50HZ FARRULC190-SAC50HZ FARRULC190-SWC50HZ FARRULC259-DAC50HZ FARRULC259-DWC50HZ FARRULC259-SWC50HZ FARRULC259-BAC50HZ FARRULC259-SAC50HZ FARRULC328-SWC50HZ
FARRAR® ULC Ultra-Low Chambers
Pakastimet Ultra Low Temperature Freezers
Purpose-built for life science applications, FARRAR®’s ULC series including the Ultra-Low Chamber (ULC-190), Ultra-Low Chamber (ULC-328), and the new Ultra-Low Chamber (ULC-259) are the only −20 to −80 °C, forced-air, ultra-low-temperature freezers on the market with air and water-cooled options.
  • Optimized footprint - 50% less floorspace versus standard ULT configurations with six times the capacity (one ULC equals six ULTs)
  • Industry's fastest temperature recovery - 10× faster temperature recovery from door opening helps ensure product integrity
  • Integrated,