Master mixes, hot start, Phusion® U

Toimittaja: FINNZYMES REAGENTS

F-533L F-533S
F-533LEA 1130 EUR
F-533L F-533S
Master mixes, hot start, Phusion® U
Nukleiinihapporeagenssit End-point PCR Enzymes and Kits

Phusion® U hot start PCR master mix is a novel engineered high fidelity enzyme developed using fusion technology.


  • Accuracy: high fidelity DNA polymerase (25X Taq)
  • Uracil-tolerance: engineered to incorporate dUTP and amplify uracil containing templates
  • Specificity: hot start for reduced nonspecific amplification and primer degradation
  • Speed: short extension times (15 to 30 s/kb)
  • Store at –20 °C


Due to a proprietary mutation in the so called dUTP binding pocket of Phusion®, Phusion® U overcomes an important limitation of proofreading enzymes - it can incorporate dUTP and read through uracil present in DNA templates.


In addition to uracil usage possibility, it features all the superior properties of other Phusion® DNA polymerases - great accuracy, speed, ability to amplify long amplicons up to 20 kb and a high specificity with Affibody based hot start. These features make DNA Polymerase an excellent choice for such important applications as amplification of bisulphite converted or damaged DNA as well as use of carryover contamination control.


It is convenient 2X mix containing Phusion® U hot start DNA polymerase, nucleotides and optimised reaction buffer including MgCl₂. Only template and primers need to be added by the user.


Toimitustiedot: Includes: Phusion® U hot start DNA polymerase, 2X reaction buffer, 3 mm MgCl₂, 400 µm of each dATP, dTTP, dCTP and dGTP. The master mix is supplied with nuclease-free water.


Varoitus: For research use only. Not for use in diagnostic procedures.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR