Anti-BACH1 Goat Polyclonal Antibody

Toimittaja: Everest Biotech
EB12081
EVBIEB12081EA 335 EUR
EVBIEB12081
Anti-BACH1 Goat Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: BACH1 (BTB and CNC homology 1, basic leucine zipper transcription factor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR