Anti-BOD1L1 Goat Polyclonal Antibody

Toimittaja: Everest Biotech
EB11986
EVBIEB11986EA 335 EUR
EVBIEB11986
Anti-BOD1L1 Goat Polyclonal Antibody
Western Blot: Not yet tested - our routinely used western blotting protocol does not allow detection of proteins as large as the calculated size of 330kDa according to NP_683692.2. Therefore we cannot recommend an optimal concentration and the antibody i

Type: Primary
Antigen: BOD1L1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR