Anti-MNSOD Goat Polyclonal Antibody

Toimittaja: Everest Biotech
EB10972
EVBIEB10972EA 335 EUR
EVBIEB10972
Anti-MNSOD Goat Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: MNSOD
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR