Anti-PCBP1 Goat Polyclonal Antibody

Toimittaja: Everest Biotech
EB10498
EVBIEB10498EA 335 EUR
EVBIEB10498
Anti-PCBP1 Goat Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: PCBP1 (poly(rC) binding protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR