Anti-LMTK2 Goat Polyclonal Antibody

Toimittaja: Everest Biotech
EB10304
EVBIEB10304EA 335 EUR
EVBIEB10304
Anti-LMTK2 Goat Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: LMTK2 (lemur tyrosine kinase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Dog, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR