Anti-Cryopirin / NALP3 / NLRP3 Goat polyclonal antibody unconjugated

Toimittaja: Everest Biotech
EB09884
EVBIEB09884EA 335 EUR
EVBIEB09884
Anti-Cryopirin / NALP3 / NLRP3 Goat polyclonal antibody unconjugated
Vasta-aineet
Type: Primary
Antigen: Cryopirin / NALP3 / NLRP3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR