Anti-GPI / Neuroleukin Goat Polyclonal Antibody

Toimittaja: Everest Biotech
EB09739
EVBIEB09739EA 335 EUR
EVBIEB09739
Anti-GPI / Neuroleukin Goat Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: GPI / Neuroleukin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Human, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR