Anti-MARK2 Goat Polyclonal Antibody

Toimittaja: Everest Biotech
EB08426
EVBIEB08426EA 335 EUR
EVBIEB08426
Anti-MARK2 Goat Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: MARK2 (MAP/microtubule affinity-regulating kinase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR