Anti-CDHR5 Goat polyclonal antibody unconjugated

Toimittaja: Everest Biotech
EB08372
EVBIEB08372EA 194 EUR
EVBIEB08372
Anti-CDHR5 Goat polyclonal antibody unconjugated
Vasta-aineet
Type: Primary
Antigen: CDHR5 (cadherin-related family member 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR