Anti-ADORA1 Goat Polyclonal Antibody

Toimittaja: Everest Biotech
EB08054
EVBIEB08054EA 335 EUR
EVBIEB08054
Anti-ADORA1 Goat Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: ADORA1 (adenosine A1 receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR