Anti-WDFY1 Goat Polyclonal Antibody

Toimittaja: Everest Biotech
EB06341
EVBIEB06341EA 194 EUR
EVBIEB06341
Anti-WDFY1 Goat Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: WDFY1 (WD repeat and FYVE domain containing 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR