Anti-MUTYH Goat Polyclonal Antibody

Toimittaja: Everest Biotech
EB06329
EVBIEB06329EA 335 EUR
EVBIEB06329
Anti-MUTYH Goat Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: MUTYH (mutY homolog)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR