Anti-DDAH1 Goat polyclonal antibody unconjugated

Toimittaja: Everest Biotech
EB05278
EVBIEB05278EA 335 EUR
EVBIEB05278
Anti-DDAH1 Goat polyclonal antibody unconjugated
Vasta-aineet
Type: Primary
Antigen: DDAH1 (dimethylarginine dimethylaminohydrolase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: Bovine, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR