Ion selective meter, bench top, Eutech ION 2700
Toimittaja: EUTECH
ECION270040S ECION270042GS
EUTEECION270040SEA 1210 EUR
EUTEECION270040S EUTEECION270042GS
Ion selective meter, bench top, Eutech ION 2700
Sähkökemiamittarit Ionimittarit
This bench meter comes with a large, one-glance-sees-all screen. Users can view pH, ION or Redox readings together with temperature, electrode status, calibration points, date and time all at once.
  • Up to 6-point push button calibration with auto-buffer recognition
  • Quick, easy electrode diagnosis with pH slope and offset display
  • Non volatile memory holds up to 500 data points – time and date-stamped for GLP compliance
  • Bi-directional RS232 for easy data transfer to computer
  • Cal-due alarm – no more out-dated calibrations
  • Auto-logging function for convenient