Lämpötila- ja kosteusloggeri, iMINI
Toimittaja: Askey Dataloggers
MX-OE-S-8-L
ESCOMX-OE-S-8-LEA 130 EUR
ESCOMX-OE-S-8-L
Lämpötila- ja kosteusloggeri, iMINI
Dataloggerit
The iMINI data loggers are designed for the monitoring of both temperature and humidity on sensitive products. They are available as disposable or reusable loggers. They encompass a full spectrum of uses from food processing and warehousing, to blood storage, transport and clinical trials.
  • Visual alarms: Two red and two blue LED
  • Large LCD display with start and stop buttons
  • NIST Traceable® calibration and bookmarking capability
Sertifikaatit: ISO 9001:2008, CE, RoHS compliant; accuracy certified.