Watchmakers' magnifiers
Toimittaja: ESCHENBACH OPTIK
1130 11245 11247 1124110
ESCH1130EA 90.2 EUR
ESCH1130 ESCH11245 ESCH11247 ESCH1124110
Watchmakers' magnifiers
Suurennuslasit
PXM® lightweight lenses with antistatic and scratch resistant coating. Ideal as a watchmaker's magnifier or gemstone magnifier.
  • Includes vent to prevent condensation
  • Made of flame resistant plastic material
  • Colour: anthracite