Lab air quality sensor, Halo Sense
Toimittaja: ERLAB CAPTAIRE
AS016010021 AS036020021 AS046030022
ERLAAS016010021EA 1390 EUR
ERLAAS016010021 ERLAAS036020021 ERLAAS046030022
Lab air quality sensor, Halo Sense
Valvontajärjestelmät Environmental Monitoring Systems
Halo Sense has been designed to continuously monitor the lab air quality for a large spectrum of fumes and vapours. Halo Sense comes in 3 versions: Volatile Organic Compounds (VOC), acid and formaldehyde.
Please complete the quote request to enable us to assist you in finding the right unit for your application.
https://fi.cmd2.vwr.com/bin/public/fc/main?f=Y5&pimid=20525804
  • Simple communication by LED pulsation system
  • Power supplies in compliance with various international standards
  • RJ45 cable provided
  • Default settings: audible alarm disabled, visual alarm activated (both alarms can