Bumper Guards
Toimittaja: ERGOMAT
HCPB100-BK HCPB100-ESD-BK HCIB120 SIBP120 LHCPB120 RCB120-ESD-BK XLSCB120 LRSB120-BK RSB120-ESD-BK
ERGOHCPB100-BKEA 39.9 EUR
ERGOHCPB100-BK ERGOHCPB100-ESD-BK ERGOHCIB120 ERGOSIBP120 ERGOLHCPB120 ERGORCB120-ESD-BK ERGOXLSCB120 ERGOLRSB120-BK ERGORSB120-ESD-BK
Bumper Guards
Kalusteet Latches and Bump Protection
These Ergomat bumper guards, cart stops and edge protectors offer a variety of devices to prevent personal injury or product damage in a dynamic work environment.
  • Resistant polyurethane foam with self-adhesive attachment to rack beams, posts and uprights
  • Use anywhere workers might run into objects - supports, stands, low hanging objects
  • Round, flat, and edge surface protection
  • Yellow and black hazard striped
These guards prevent cuts and bruises from sharp edges with edge protecting bumper guards that slide over the sharp edge and holdMore Product Information