Cleanroom anti-fatigue mats, Ergomat Nitro
Toimittaja: ERGOMAT
ENT0203 ENT0420 ENT0412 ENT0406 ENT0320 ENT0306 ENT0304 ENT0220 ENT0206 ENT0209 ENT0212 ENT0204 ENT0303 ENT0309 ENT0312 ENT0404 ENT0409
115-0895EA 153 EUR
115-0895 115-0916 115-0915 115-0914 115-0913 115-0912 115-0899 115-0898 115-0896 115-0897 ERGOENT0212 ERGOENT0204 ERGOENT0303 ERGOENT0309 ERGOENT0312 ERGOENT0404 ERGOENT0409
Cleanroom anti-fatigue mats, Ergomat Nitro
Matot Ergonomiset työtilamatot
Ergomat® Nitro with anti-slip cone profile is made from pure nitrile rubber and is designed to increase comfort while reducing trip hazards, body pain, quality issues, and sick leave. It is ideal for usage in the electronics industry or for areas where static control is crucial due to sensitive equipment.
  • Upper side with cones
  • Autoclavable, withstands 600 °C (welding)
  • Chemical exposure will not affect the state of the mat; no curling up, swelling, or expanding
  • Moulded bevelled edges prevent curling, eliminates trip hazards
  • Raw virgin material guarantee