Signs, floor signs, DuraStripe®, door swing
Toimittaja: ERGOMAT
4070089AP53 4070090AP71 4070091AP86 4070092AP55 4070093AP77 4070094AP88
ERGO4070089AP53EA 116 EUR
ERGO4070089AP53 ERGO4070090AP71 ERGO4070091AP86 ERGO4070092AP55 ERGO4070093AP77 ERGO4070094AP88
Signs, floor signs, DuraStripe®, door swing
Merkit, kyltit ja lattiamerkinnät
DuraStripe corners are available in an array of die-cut peel-&-stick colours. These durable floor markings will communicate safety instructions and can withstand pedestrian or forklift traffic. DuraStripe corners are so easy to install and reconfigure, you just peel-&-stick! This will save you time and money from having to paint floor markings.
  • Ideal for perfect or less than perfect floors
  • Simple and easy peel-&-stick installation
  • No dry time, cure time, fumes or messy clean up to limit productivity