Restriction enzymes, NotI, 10 U/µl, Fermentas
Toimittaja: Thermo Fisher Scientific
ER0591 ER0592 ER0595
ER0591EA 73.5 EUR
ER0591 ER0592 ER0595
Restriction enzymes, NotI, 10 U/µl, Fermentas
Nukleiinihapporeagenssit Restriction Enzymes
NotI restriction enzyme recognises GC^GGCCGC sites and cuts best at 37 °C in O buffer.
  • Superior quality, stringent quality control and industry leading manufacturing process
  • Convenient colour-coded five buffer system
  • Includes universal Tango buffer for double-digestions
  • BSA premixed in reaction buffers
  • Wide selection of restriction endonuclease specificities
Applications: Molecular cloning, restriction site mapping, genotyping, southern blotting, restriction fragment length polymorphism (RFLP), SNP.