Purkki, leveä suuaukko, avonaisella korkilla
Toimittaja: EP SCIENTIFIC
S320-0060 S321-0250 S340-0120 S120-0125
EPSCS320-0060EA 256 EUR
EPSCS320-0060 EPSCS321-0250 EPSCS340-0120 EPSCS120-0125
Purkki, leveä suuaukko, avonaisella korkilla
Astiat ja purkit Astiat, yleiskäyttö
Type III soda-lime glass
  • Designed for water sample collection for volatile organic analysis (VOA) and soil samples
  • Caps assembled along with bottles
  • Amber jars are recommended for light-sensitive analytes
  • With open-top white PP cap bonded with PTFE-lined silicone septum
Processed or certified to meet or exceed EPA Performance Based Specifications for volatile organic analysis with certificate. Processed vials are processed in the same manner as certified jars and come with all the features of certified jars, but jars are not barcoded and CofA is not included.