Transferrin ELISA kit, Enzo Life Sciences
Toimittaja: ENZO LIFE SCIENCES
ENZKIT1430001
ENZOENZKIT1430001EA 757 EUR
ENZOENZKIT1430001
Transferrin ELISA kit, Enzo Life Sciences
Bio-assayt ELISA-kitit
Highly sensitive Transferrin ELISA kit enabling detection of transferrin in serum and plasma in just 2 hours.
  • Sensitive measurement of transferrin, detecting as little as 4,6 ng/ml
  • Negligible cross reactivity with similar proteins
  • High throughput format with results in 2 hours for up to 40 samples in duplicate
  • Fully quantitative results that surpass semi-quantitative Western blot analysis
For the quantitative determination of transferrin in human serum, human plasma and canine serum. The Transferrin ELISA kit is a colorimetric competitive immunoassay kit with results in 2 hours.
Tilaustiedot: Contains Microtiter Plate, Assay Buffer 13, Standard, Wash Buffer Concentrate, Antibody, Antibody Diluent, Tracer, Conjugate, Substrate, and Stop Solution.