ApoE ELISA kit, grow cells
Toimittaja: ENZO LIFE SCIENCES
ENZKIT1340001
ENZOENZKIT1340001EA 901 EUR
ENZOENZKIT1340001
ApoE ELISA kit, grow cells
Bio-assayt ELISA-kitit
Highly sensitive ApoE ELISA kit enabling detection of ApoE in serum, plasma, cell culture supernates, and tissue homogenates in 3,5 hours.
  • Sensitive measurement of ApoE, detecting as little as 14,063 pg/ml
  • Negligible reactivity to similar proteins.
  • High throughput with results in 3,5 hours.
  • Fully quantitative results that surpass semi-quantitative Western blot analysis
For the quantitative determination of ApoE in serum, plasma, cell culture supernates and tissue homogenates in human samples.
The ApoE ELISA kit is a colorimetric immunometric immunoassay kit with results in 3,5 hrs.
Tilaustiedot: Contains Microtiter Plate, Standard, Sample Buffer, Wash Buffer Concentrate, Antibody, Antibody Buffer, Conjugate, Conjugate Buffer Substrate, and Stop Solution.