JK 184 ≥99% (by HPLC)

Toimittaja: ENZO LIFE SCIENCES

ENZCHM1290005
ENZOENZCHM1290005EA 70.4 EUR
ENZOENZCHM1290005
JK 184 ≥99% (by HPLC)
JK 184

Hedgehog (Hh) pathway blocker.

Kaava: C₁₉H₁₈N₄OS
MDL numero: MFCD01044465
CAS-numero: 315703-52-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=75d795eae651e3f3fafb6d2c755ea8e9e004bacd&vid=d42b1236472411fb119f45cd4c1c2b551ec04da8&prid=web&clid=SAPDAM

Order Now

TEKNISET TIEDOT

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR