DNA isolation kit, EPIXTRACT®
Toimittaja: ENZO LIFE SCIENCES
ENZ450180050 ENZ450180100
ENZOENZ450180050EA 247 EUR
ENZOENZ450180050 ENZOENZ450180100
DNA isolation kit, EPIXTRACT®
Nukleiinihapporeagenssit Nucleic Acid Purification Kits and Reagents
EPIXTRACT® DNA isolation kit provides a simple method to isolate DNA from plasma, serum, and body fluids.
  • Rapid, provide results in less than 15 minutes
  • Safe to use, contains only non-toxic reagents and no phenol chloroform
  • Highly efficient, isolate DNA from plasma or serum containing as low as 0,1 ng DNA
Toimitustiedot: Supplied with digestion solution, digestion powder, DNA isolation buffer, DNA elution solution, spin column and collection tube.